Anti-CD34 Mouse Monoclonal Antibody (CF660R) [clone: QBEnd/10]
Toimittaja: Biotium
BNC610640-500 BNC610640-100
BNC610640-500EA 560 EUR
BNC610640-500 BNC610640-100
Anti-CD34 Mouse Monoclonal Antibody (CF660R) [clone: QBEnd/10]
Vasta-aineet
This MAb recognizes a single chain, transmembrane, heavily glycosylated protein of 90-120 kDa, which is identified as CD34. On the basis of differential sensitivity to degradation by specific enzymes, epitopes of monoclonal antibodies to CD34 are classified intothree main categories, class I, class II and class III. It is a class II antibody whose epitope is resistant to neuraminidase but sensitive to glycoprotease and chymopapain.CD34 expression is a hallmark for identifying pluripotent hematopoietic stem or progenitor cells. Its expression is gradually lost as lineage committed progenitorsMore Product Information