Anti-CD33 Mouse Monoclonal Antibody (CF647) [clone: C33/68]
Toimittaja: Biotium
BNC470068-500 BNC470068-100
BNC470068-500EA 560 EUR
BNC470068-500 BNC470068-100
Anti-CD33 Mouse Monoclonal Antibody (CF647) [clone: C33/68]
Vasta-aineet
Recognizes a 67 kDa glycoprotein, which is identified as CD33. It is a transmembrane protein of the sialic acid-binding immunoglobulin-like lectin (Siglec) family. It belongs to the immunoreceptor tyrosine-based inhibitory motif (ITIM)-containing molecules able of recruiting protein tyrosine phosphatases SHP-1 and SHP-2 to signal assemblies; these ITIMs are also used for ubiquitin-mediated removal of the receptor from the cell surface. CD33 is expressed on cells of myelomonocytic lineage, binds sialic acid residues in N- and O-glycans on cell surfaces, and is a therapeutic target for acuteMore Product Information