Anti-Plasma Cell Marker Mouse Monoclonal Antibody (CF647) [clone: 7B18]
Toimittaja: Biotium
BNC470047-500 BNC470047-100
BNC470047-500EA 560 EUR
BNC470047-500 BNC470047-100
Anti-Plasma Cell Marker Mouse Monoclonal Antibody (CF647) [clone: 7B18]
Vasta-aineet
This antibody recognizes an intra-cytoplasmic antigen, which shows a very high degree of specificity for plasma cells. This antigen is present in normal as well as neoplastic plasma cells. Plasma cells, which are large lymphocytes derived from an antigen-specific B cell, secrete antibodies and are responsible for humoral immunity. Plasma cells differentiate from B cells upon stimulation by CD4 lymphocytes. The B cell acts as an antigen-presenting cell (APC), consuming an offending pathogen, which is taken up by the B cell by phagocytosis and broken down within proteosomes. Plasma cellsMore Product Information