Anti-MYOG Mouse Monoclonal Antibody (CF640R) [clone: MGN185 F5D]
Toimittaja: Biotium
BNC401233-500 BNC401233-100
BNC401233-500EA 560 EUR
BNC401233-500 BNC401233-100
Anti-MYOG Mouse Monoclonal Antibody (CF640R) [clone: MGN185 F5D]
Vasta-aineet
Myogenin is a member of the MyoD family of myogenic basic helix-loop-helix (bHLH) transcription factors that also includes MyoD, Myf-5, and MRF4 (also known as herculinor Myf-6). MyoD family members are expressed exclusively in skeletal muscle and play a key role in activating myogenesis by binding to enhancer sequences of muscle-specific genes. The regulatory domain of MyoD is approximately 70 amino acids in length and includes both a basic DNA binding motif and a bHLH dimerization motif. MyoD family members share about 80% amino acid homology in their bHLH motifs. Anti-myogenin labels theMore Product Information