Anti-SUMO2 Mouse Monoclonal Antibody (CF640R) [clone: SUMO2/1199]
Toimittaja: Biotium
BNC401199-500 BNC401199-100
BNC401199-500EA 560 EUR
BNC401199-500 BNC401199-100
Anti-SUMO2 Mouse Monoclonal Antibody (CF640R) [clone: SUMO2/1199]
Vasta-aineet
The small ubiquitin-related modifier (SUMO) proteins, which include SUMO-1, 2 and 3, belong to the ubiquitin-like protein family. Like ubiquitin, the SUMO proteins are synthesized as precursor proteins that undergo processing before conjugation to target proteins. Also, both utilize the E1, E2 and E3 cascade enzymes for conjugation. However, SUMO and ubiquitin differ with respect to targeting. Ubiquitination predominantly targets proteins for degradation, whereas sumoylation targets proteins to a variety of cellular processing, including nuclear transport, transcriptional regulation,More Product Information