Anti-PECAM1 Mouse Monoclonal Antibody (CF640R) [clone: C31.3 C31.7 C31.10]
Toimittaja: Biotium
BNC401099-500 BNC401099-100
BNC401099-500EA 560 EUR
BNC401099-500 BNC401099-100
Anti-PECAM1 Mouse Monoclonal Antibody (CF640R) [clone: C31.3 C31.7 C31.10]
Vasta-aineet
CD31 (PECAM-1) is a transmembrane glycoprotein member of the immunoglobulin supergene family of adhesion molecules. CD31 is expressed by stem cells of the hematopoietic system and is primarily used to identify and concentrate these cells for experimental studies as well as for bone marrow transplantation. Anti-CD31 has shown to be highly specific and sensitive for vascular endothelial cells. Staining of nonvascular tumors (excluding hematopoietic neoplasms) is rare. CD31 MAb reacts with normal, benign, and malignant endothelial cells which make up blood vessel lining. The level of CD31More Product Information