Anti-THY1 Mouse Monoclonal Antibody (CF640R) [clone: F15-42-1]
Toimittaja: Biotium
BNC400194-500 BNC400194-100
BNC400194-500EA 560 EUR
BNC400194-500 BNC400194-100
Anti-THY1 Mouse Monoclonal Antibody (CF640R) [clone: F15-42-1]
Vasta-aineet
CD90 (Thy-1) is an 18-35 kDa GPI-anchored plasma membrane glycoprotein expressed in many cell types, such as in hematopoietic cells and neurons, connective tissues, various fibroblast and stromal cell lines, tumor endothelial cell lines and others. It is involved in T-cell activation, cellular adhesion, proliferation and migration, neurite outgrowth, wound healing, apoptosis, and fibrosis. CD90 participates in multiple signaling cascades and its effects are tissue- and cell type-specific. It often functions as an important regulator of cell-cell and cell-matrix interactions.
Type: Primary