Anti-THY1 Mouse Monoclonal Antibody (CF405M) [clone: AF-9]
Toimittaja: Biotium
BNC050987-500 BNC050987-100
BNC050987-500EA 560 EUR
BNC050987-500 BNC050987-100
Anti-THY1 Mouse Monoclonal Antibody (CF405M) [clone: AF-9]
Vasta-aineet
Recognizes a protein of 18-35 kDa, identified as CD90 (also known as Thy1). CD90 is a member of the immunoglobulin superfamily. It may contribute to inhibition of proliferation/differentiation of hematopoietic stem cells and neuron memory formation in the CNS. It consists of a single Ig domain (112 amino acids; 25-35 kDa) inserted into the cell membrane via a GPI anchor. Expressed by hematopoietic stem cells and neurons in all species studied. Its highly expressed in connective tissue and various fibroblast and stromal cell lines, expressed on all thymocytes and peripheral T cells in mice,More Product Information