Kuoppalevyt, syväkuoppalevyt, 96-kuoppaiset nelikulmaiset
Toimittaja: BIOTIX
63300108 63300109
732-2482EA 37.6 EUR
732-2482 BITX63300109
Kuoppalevyt, syväkuoppalevyt, 96-kuoppaiset nelikulmaiset
Kuoppalevyt Syväkuoppalevyt
Medical grade virgin PP, non treated. These sturdy 96-deep well microplates conform to SBS standards for automated high throughput processing and compound storage.
  • Alphanumeric grid to help with sample identification; notched bottom right corner to ensure correct plate orientation
  • Conical shaped wells facilitate sample recovery
  • Each plate can withstand centrifugation up to 6000 RCF when supported by the appropriate microplate rotor
  • Excellent chemical resistance and temperature resistant from +121 to −80 °C
  • Stackable to save space in incubators and freezers