Anti-DLL3 Rabbit polyclonal antibody unconjugated

Toimittaja: Biorbyt
ORB97038-100
BIRBORB97038-100EA 0 EUR
BIRBORB97038-100
Anti-DLL3 Rabbit polyclonal antibody unconjugated
Vasta-aineet
Type: Primary
Antigen: DLL3 (delta-like 3 (Drosophila))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR