Anti-Rho Kinase Rabbit Polyclonal Antibody

Toimittaja: Biorbyt
ORB96202-50
BIRBORB96202-50EA 0 EUR
BIRBORB96202-50
Anti-Rho Kinase Rabbit Polyclonal Antibody
Vasta-aineet
Immunochemistry: 1:200; Western Blot: 1:2000

Type: Primary
Antigen: Rho Kinase
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR