Anti-NPY5R Goat Polyclonal Antibody

Toimittaja: Biorbyt
ORB94932-50
BIRBORB94932-50EA 0 EUR
BIRBORB94932-50
Anti-NPY5R Goat Polyclonal Antibody
Vasta-aineet
ELISA: 1:32000; Immunochemistry: 3.75 ?g/ml; Western Blot: 0.3-1 ?g/ml

Type: Primary
Antigen: NPY5R (neuropeptide Y receptor Y5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR