Anti-STAT1 Rabbit Polyclonal Antibody

Toimittaja: Biorbyt
ORB94850-50
BIRBORB94850-50EA 0 EUR
BIRBORB94850-50
Anti-STAT1 Rabbit Polyclonal Antibody
Vasta-aineet
ELISA: 1:10000; Immunochemistry: 10 ?g/ml; Western Blot: 1:500-1:1000

Type: Primary
Antigen: STAT1 (signal transducer and activator of transcription 1, 91kDa)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR