Anti-FNBP1 Goat Polyclonal Antibody

Toimittaja: Biorbyt
ORB94075-50
BIRBORB94075-50EA 0 EUR
BIRBORB94075-50
Anti-FNBP1 Goat Polyclonal Antibody
Vasta-aineet
ELISA: 1:32000; Immunochemistry: 3.75 ?g/ml; Western Blot: 1:32000, 0.25-2 ?g/ml

Type: Primary
Antigen: FNBP1 (formin binding protein 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR