Anti-NPHS1 Rabbit Polyclonal Antibody

Toimittaja: Biorbyt
ORB89904-50
BIRBORB89904-50EA 0 EUR
BIRBORB89904-50
Anti-NPHS1 Rabbit Polyclonal Antibody
Vasta-aineet
Immunochemistry: 5 ?g/ml

Type: Primary
Antigen: NPHS1 (nephrosis 1, congenital, Finnish type (nephrin))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR