Anti-YWHAZ Rabbit Polyclonal Antibody

Toimittaja: Biorbyt
ORB87257-50
BIRBORB87257-50EA 0 EUR
BIRBORB87257-50
Anti-YWHAZ Rabbit Polyclonal Antibody
Vasta-aineet
Immunochemistry: 10 ?g/ml

Type: Primary
Antigen: YWHAZ (tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, zeta)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR