Anti-Transmissible Gastroenteritis Pig Virus Mouse Monoclonal Antibody

Toimittaja: Biorbyt
ORB79503-1
BIRBORB79503-1EA 0 EUR
BIRBORB79503-1
Anti-Transmissible Gastroenteritis Pig Virus Mouse Monoclonal Antibody
Vasta-aineet
Type: Primary
Antigen: Transmissible Gastroenteritis Pig Virus
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR