Anti-EFCAB14 Rabbit polyclonal antibody unconjugated

Toimittaja: Biorbyt
ORB78210-100
BIRBORB78210-100EA 0 EUR
BIRBORB78210-100
Anti-EFCAB14 Rabbit polyclonal antibody unconjugated
Vasta-aineet
ELISA: 1:20000-1:80000; Western Blot: 1:500-1:1000

Type: Primary
Antigen: EFCAB14 (EF-hand calcium binding domain 14)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR