Anti-ABCB5 Rabbit Polyclonal Antibody

Toimittaja: Biorbyt
ORB77741-100
BIRBORB77741-100EA 0 EUR
BIRBORB77741-100
Anti-ABCB5 Rabbit Polyclonal Antibody
Vasta-aineet
ELISA: 1:20000-1:80000; Western Blot: 1:500-1:1000

Type: Primary
Antigen: ABCB5 (ATP-binding cassette, sub-family B (MDR/TAP), member 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR