Anti-ADAM19 Rabbit Polyclonal Antibody

Toimittaja: Biorbyt
ORB77011-100
BIRBORB77011-100EA 0 EUR
BIRBORB77011-100
Anti-ADAM19 Rabbit Polyclonal Antibody
Vasta-aineet
ELISA: 1:10000-1:40000; Western Blot: 1:200-1:500

Type: Primary
Antigen: ADAM19 (ADAM metallopeptidase domain 19)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR