Anti-GZMB Rabbit Polyclonal Antibody

Toimittaja: Biorbyt
ORB76251-100
BIRBORB76251-100EA 0 EUR
BIRBORB76251-100
Anti-GZMB Rabbit Polyclonal Antibody
Vasta-aineet
Type: Primary
Antigen: GZMB (granzyme B (granzyme 2, cytotoxic T-lymphocyte-associated serine esterase 1))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR