Anti-SLITRK2 Rabbit Polyclonal Antibody

Toimittaja: Biorbyt
ORB74936-100
BIRBORB74936-100EA 0 EUR
BIRBORB74936-100
Anti-SLITRK2 Rabbit Polyclonal Antibody
Vasta-aineet
Type: Primary
Antigen: SLITRK2 (SLIT and NTRK-like family, member 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR