Anti-PRPSAP2 Rabbit Polyclonal Antibody

Toimittaja: Biorbyt
ORB69942-100
BIRBORB69942-100EA 0 EUR
BIRBORB69942-100
Anti-PRPSAP2 Rabbit Polyclonal Antibody
Vasta-aineet
Type: Primary
Antigen: PRPSAP2 (phosphoribosyl pyrophosphate synthetase-associated protein 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR