Anti-SHC4 Rabbit Polyclonal Antibody

Toimittaja: Biorbyt
ORB69939-100
BIRBORB69939-100EA 0 EUR
BIRBORB69939-100
Anti-SHC4 Rabbit Polyclonal Antibody
Vasta-aineet
Type: Primary
Antigen: SHC4 (SHC (Src homology 2 domain containing) family, member 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR