Anti-KIT Mouse Monoclonal Antibody (FITC (Fluorescein))

Toimittaja: Biorbyt
ORB44629-100
BIRBORB44629-100EA 0 EUR
BIRBORB44629-100
Anti-KIT Mouse Monoclonal Antibody (FITC (Fluorescein))
Vasta-aineet
Type: Primary
Antigen: KIT (v-kit Hardy-Zuckerman 4 feline sarcoma viral oncogene homolog)
Clonality: monoclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR