Anti-CD7 Mouse Monoclonal Antibody (Peridinin Chlorophyll)

Toimittaja: Biorbyt
ORB44609-100
BIRBORB44609-100EA 0 EUR
BIRBORB44609-100
Anti-CD7 Mouse Monoclonal Antibody (Peridinin Chlorophyll)
Vasta-aineet
Type: Primary
Antigen: CD7 (CD7 molecule)
Clonality: monoclonal
Clone:
Conjugation: Peridinin Chlorophyll
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR