Anti-MAN1B1 Rabbit Polyclonal Antibody

Toimittaja: Biorbyt
ORB41199-100
BIRBORB41199-100EA 0 EUR
BIRBORB41199-100
Anti-MAN1B1 Rabbit Polyclonal Antibody
Vasta-aineet
Western Blot: 1:500-1:3000; Immunochemistry: 1:100-1:250

Type: Primary
Antigen: MAN1B1 (mannosidase, alpha, class 1B, member 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR