Anti-PDI Rabbit Polyclonal Antibody

Toimittaja: Biorbyt
ORB40838-100
BIRBORB40838-100EA 0 EUR
BIRBORB40838-100
Anti-PDI Rabbit Polyclonal Antibody
Vasta-aineet
Western Blot: 1:500-1:3000

Type: Primary
Antigen: PDI
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR