Anti-CASKIN2 Rabbit polyclonal antibody

Toimittaja: Biorbyt
ORB2702-100 ORB2702-200
BIRBORB2702-100EA 0 EUR
BIRBORB2702-100 BIRBORB2702-200
Anti-CASKIN2 Rabbit polyclonal antibody
Vasta-aineet
Western Blot: 1:100-500; Immunochemistry: 1:50-250 (based on 0.5 mg/ml)

Type: Primary
Antigen: CASKIN2 (CASK interacting protein 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR