Anti-BrdU Mouse monoclonal antibody

Toimittaja: Biorbyt
ORB26152-100
BIRBORB26152-100EA 0 EUR
BIRBORB26152-100
Anti-BrdU Mouse monoclonal antibody
Vasta-aineet
Type: Primary
Antigen: BrdU (Bromodeoxyuridine)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR