Anti-CCDC38 Rabbit Polyclonal Antibody

Toimittaja: Biorbyt
ORB2521-100 ORB2521-200
BIRBORB2521-100EA 0 EUR
BIRBORB2521-100 BIRBORB2521-200
Anti-CCDC38 Rabbit Polyclonal Antibody
Vasta-aineet
Western Blot: 1:100-500; Immunochemistry: 1:50-250 (based on 0.5 mg/ml)

Type: Primary
Antigen: CCDC38 (coiled-coil domain containing 38)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR