Anti-SCC112 Rabbit Polyclonal Antibody

Toimittaja: Biorbyt
ORB2346-100 ORB2346-200
BIRBORB2346-100EA 0 EUR
BIRBORB2346-100 BIRBORB2346-200
Anti-SCC112 Rabbit Polyclonal Antibody
Vasta-aineet
Western Blot: 1:100-500; Immunochemistry: 1:50-250 (based on 0.5 mg/ml)

Type: Primary
Antigen: SCC112
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR