Anti-Influenza B HA2 Mouse Monoclonal Antibody

Toimittaja: Biorbyt
ORB23355-1
BIRBORB23355-1EA 0 EUR
BIRBORB23355-1
Anti-Influenza B HA2 Mouse Monoclonal Antibody
Vasta-aineet
Type: Primary
Antigen: Influenza B HA2
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR