Anti-IgG Rabbit Polyclonal Antibody

Toimittaja: Biorbyt
ORB22313-1
BIRBORB22313-1EA 0 EUR
BIRBORB22313-1
Anti-IgG Rabbit Polyclonal Antibody
Vasta-aineet
Type: Secondary
Antigen: IgG (Duck) (duck IgG)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR