Anti-UOX Rabbit Polyclonal Antibody (Biotin)

Toimittaja: Biorbyt
ORB21092-1
BIRBORB21092-1EA 0 EUR
BIRBORB21092-1
Anti-UOX Rabbit Polyclonal Antibody (Biotin)
Vasta-aineet
Type: Primary
Antigen: UOX (urate oxidase, pseudogene)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR