Anti-HOXD13 Goat Polyclonal Antibody

Toimittaja: Biorbyt
ORB20481-100
BIRBORB20481-100EA 0 EUR
BIRBORB20481-100
Anti-HOXD13 Goat Polyclonal Antibody
Vasta-aineet
ELISA: 1:8000; Western Blot: 1 ?g/ml

Type: Primary
Antigen: HOXD13 (homeobox D13)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR