Anti-TGFBI Goat Polyclonal Antibody

Toimittaja: Biorbyt
ORB20453-100
BIRBORB20453-100EA 0 EUR
BIRBORB20453-100
Anti-TGFBI Goat Polyclonal Antibody
Vasta-aineet
ELISA: 1:32000

Type: Primary
Antigen: TGFBI (transforming growth factor, beta-induced, 68kDa)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR