Anti-CRCT1 Rabbit polyclonal antibody

Toimittaja: Biorbyt
ORB1972-100 ORB1972-200
BIRBORB1972-100EA 0 EUR
BIRBORB1972-100 BIRBORB1972-200
Anti-CRCT1 Rabbit polyclonal antibody
Vasta-aineet
Western Blot: 1:100-500; Immunochemistry: 1:50-250 (based on 0.5 mg/ml)

Type: Primary
Antigen: CRCT1 (cysteine-rich C-terminal 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR