Anti-CCL3 and CCL3L1 Goat Polyclonal Antibody

Toimittaja: Biorbyt
ORB19059-100
BIRBORB19059-100EA 0 EUR
BIRBORB19059-100
Anti-CCL3 and CCL3L1 Goat Polyclonal Antibody
Vasta-aineet
ELISA: 1:32000; Western Blot: 1 ?g/ml

Type: Primary
Antigen: CCL3 and CCL3L1
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR