Anti-FTCD Goat Polyclonal Antibody

Toimittaja: Biorbyt
ORB18848-100 ORB18848-20
BIRBORB18848-100EA 0 EUR
BIRBORB18848-100 BIRBORB18848-20
Anti-FTCD Goat Polyclonal Antibody
Vasta-aineet
Type: Primary
Antigen: FTCD (formimidoyltransferase cyclodeaminase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR