Anti-DNM1 Rabbit Polyclonal Antibody

Toimittaja: Biorbyt
ORB135392-100 ORB135392-200 ORB135392-50
BIRBORB135392-100EA 0 EUR
BIRBORB135392-100 BIRBORB135392-200 BIRBORB135392-50
Anti-DNM1 Rabbit Polyclonal Antibody
Vasta-aineet
Type: Primary
Antigen: DNM1 (dynamin 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR