Anti-SELE Rabbit Polyclonal Antibody

Toimittaja: Biorbyt
ORB13115-100
BIRBORB13115-100EA 0 EUR
BIRBORB13115-100
Anti-SELE Rabbit Polyclonal Antibody
Vasta-aineet
Western Blot: 1:500; Immunofluorescence: 1:250

Type: Primary
Antigen: SELE (selectin E)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR