Anti-OXTR Rabbit Polyclonal Antibody (Cy5.5®)

Toimittaja: Biorbyt
ORB123325-100
BIRBORB123325-100EA 0 EUR
BIRBORB123325-100
Anti-OXTR Rabbit Polyclonal Antibody (Cy5.5®)
Vasta-aineet
Type: Primary
Antigen: OXTR (oxytocin receptor)
Clonality: polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR