Anti-CD45RA Mouse Monoclonal Antibody (PE (Phycoerythrin))

Toimittaja: Biorbyt
ORB12069-100 ORB12069-50
BIRBORB12069-100EA 0 EUR
BIRBORB12069-100 BIRBORB12069-50
Anti-CD45RA Mouse Monoclonal Antibody (PE (Phycoerythrin))
Vasta-aineet
Type: Primary
Antigen: CD45RA (CD45RA)
Clonality: monoclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR