Anti-MSH alpha Rabbit Polyclonal Antibody (Biotin)

Toimittaja: Biorbyt
ORB114323-100
BIRBORB114323-100EA 0 EUR
BIRBORB114323-100
Anti-MSH alpha Rabbit Polyclonal Antibody (Biotin)
Vasta-aineet
Type: Primary
Antigen: MSH alpha
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR