Anti-SERPINF1 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))

Toimittaja: Biorbyt
ORB113816-100
BIRBORB113816-100EA 0 EUR
BIRBORB113816-100
Anti-SERPINF1 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Vasta-aineet
Type: Primary
Antigen: SERPINF1 (serpin peptidase inhibitor, clade F (alpha-2 antiplasmin, pigment epithelium derived factor), member 1)
Clonality: polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR