Anti-Marijuana Mouse Monoclonal Antibody

Toimittaja: Biorbyt
ORB11017-100 ORB11017-200
BIRBORB11017-100EA 0 EUR
BIRBORB11017-100 BIRBORB11017-200
Anti-Marijuana Mouse Monoclonal Antibody
Vasta-aineet
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)

Type: Primary
Antigen: Marijuana
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR