Anti-Influenza B Virus Nucleoprotein Mouse Polyclonal Antibody

Toimittaja: Biorbyt
ORB10868-100 ORB10868-200
BIRBORB10868-100EA 0 EUR
BIRBORB10868-100 BIRBORB10868-200
Anti-Influenza B Virus Nucleoprotein Mouse Polyclonal Antibody
Vasta-aineet
Type: Primary
Antigen: Influenza B Virus Nucleoprotein
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR