Anti-IgG Goat Polyclonal Antibody

Toimittaja: Biorbyt
ORB108216-2
BIRBORB108216-2EA 0 EUR
BIRBORB108216-2
Anti-IgG Goat Polyclonal Antibody
Vasta-aineet
ELISA: 1:50-1:200

Type: Primary
Antigen: IgG (IgG)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR