Anti-CCL13 Rabbit polyclonal antibody

Toimittaja: Biorbyt
ORB107365-100
BIRBORB107365-100EA 0 EUR
BIRBORB107365-100
Anti-CCL13 Rabbit polyclonal antibody
Vasta-aineet
Type: Primary
Antigen: CCL13 (chemokine (C-C motif) ligand 13)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR